3-Chlor-5-(trifluormethyl)benzoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-α,α,α-trifluoro-m-toluic acid

PC6011-25G PC6011-5G PC6011-1G
APOSPC6011-25GEA 111 EUR
APOSPC6011-25G APOSPC6011-5G APOSPC6011-1G
3-Chlor-5-(trifluormethyl)benzoesäure
3-Chlor-5-(trifluormethyl)benzoesäure
Formel: C₈H₄ClF₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04115846
CAS-Nummer: 53985-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR