3-Fluor-4-iodanilin

Lieferant: Apollo Scientific
Achtung

PC5542-5G PC5542-25G
APOSPC5542-5GEA 23.2 EUR
APOSPC5542-5G APOSPC5542-25G
3-Fluor-4-iodanilin
3-Fluor-4-iodanilin
Formel: C₆H₅FIN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07774187
CAS-Nummer: 656-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR