Pentafluorbenzoylchlorid 99%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC
PC5530-100G
PC5530-25G
APOSPC5530-100GEA
122
EUR
APOSPC5530-100G
APOSPC5530-25G
Pentafluorbenzoylchlorid 99%
Pentafluorbenzoylchlorid
Useful reagent for the derivatisation of primary amines, etc for GC analysis.
Formel:
C₇ClF₅O Siedepunkt: 158…159 °C Dichte: 1.601 g/cm³ (25 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00000657 CAS-Nummer: 2251-50-5 EINECS: 218-842-4 UN-Nummer: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...