1-Brom-2-fluor-4-(trifluormethoxy)benzol
Lieferant: Apollo Scientific
|
Achtung
|
PC53215-100G
PC53215-25G
PC53215-5G
APOSPC53215-100GEA
208
EUR
APOSPC53215-100G
APOSPC53215-25G
APOSPC53215-5G
1-Brom-2-fluor-4-(trifluormethoxy)benzol
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene
Formel:
C₇H₃BrF₄O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06657548 CAS-Nummer: 168971-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...