1-[3-(Trifluoromethyl)phenyl]cyclopropanecarboxylic acid 97%

Lieferant: Apollo Scientific

PC530021-1G PC530021-5G
APOSPC530021-1GEA 140 EUR
APOSPC530021-1G APOSPC530021-5G
1-[3-(Trifluoromethyl)phenyl]cyclopropanecarboxylic acid 97%
1-[3-(Trifluoromethyl)phenyl]cyclopropanecarboxylic acid
Formel: C₁₁H₉F₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07374444
CAS-Nummer: 104173-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR