2',4'-Difluorbiphenyl-3-carbonsäure 97%
Lieferant: Apollo Scientific
PC530017-1G
PC530017-500MG
APOSPC530017-1GEA
393
EUR
APOSPC530017-1G
APOSPC530017-500MG
2',4'-Difluorbiphenyl-3-carbonsäure 97%
2',4'-Difluorbiphenyl-3-carbonsäure
Formel:
C₁₃H₈F₂O₂ Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
656304-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...