Potassium trimethoxy(trifluoromethyl)borate

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium (Trifluoromethyl)trimethoxyborate

PC52374-1G PC52374-25G PC52374-5G
APOSPC52374-1GEA 36.4 EUR
APOSPC52374-1G APOSPC52374-25G APOSPC52374-5G
Potassium trimethoxy(trifluoromethyl)borate
Potassium trimethoxy(trifluoromethyl)borate
Formel: C₄H₉BF₃KO₃
Schmelzpunkt: 116…118 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22124929
CAS-Nummer: 626232-27-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR