1-Chloro-2-(1,1-difluoroethyl)benzene 95%
Lieferant: Apollo Scientific
PC520697-10G
PC520697-1G
PC520697-5G
APOSPC520697-10GEA
838
EUR
APOSPC520697-10G
APOSPC520697-1G
APOSPC520697-5G
1-Chloro-2-(1,1-difluoroethyl)benzene 95%
1-Chloro-2-(1,1-difluoroethyl)benzene
Formel:
C₈H₇ClF₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD14525553 CAS-Nummer: 1204295-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...