5-(Trifluoromethyl)pyrimidine 95%

Lieferant: Apollo Scientific

PC520663-1G PC520663-5G
APOSPC520663-1GEA 302 EUR
APOSPC520663-1G APOSPC520663-5G
5-(Trifluoromethyl)pyrimidine 95%
5-(Trifluoromethyl)pyrimidine
Formel: C₅H₃F₃N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11100527
CAS-Nummer: 176214-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR