3-(Trifluoromethyl)-1H-indene 95%

Lieferant: Apollo Scientific

PC520649-1G PC520649-500MG PC520649-5G
APOSPC520649-1GEA 350 EUR
APOSPC520649-1G APOSPC520649-500MG APOSPC520649-5G
3-(Trifluoromethyl)-1H-indene 95%
3-(Trifluoromethyl)-1H-indene
Formel: C₁₀H₇F₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11226532
CAS-Nummer: 134391-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR