2-Bromo-4-methylbenzotrifluoride 95%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-4-methyl-1-(trifluoromethyl)benzene
PC520610-10G
PC520610-1G
PC520610-5G
APOSPC520610-10GEA
861
EUR
APOSPC520610-10G
APOSPC520610-1G
APOSPC520610-5G
2-Bromo-4-methylbenzotrifluoride 95%
2-Bromo-4-methylbenzotrifluoride
Formel:
C₈H₆BrF₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11226554 CAS-Nummer: 121793-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...