4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 98%
Lieferant: Apollo Scientific
PC520574-1G
PC520574-5G
APOSPC520574-1GEA
282
EUR
APOSPC520574-1G
APOSPC520574-5G
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid 98%
4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₃F₄NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08282778 CAS-Nummer: 878572-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...