2-(2,2,2-Trifluoroethoxy)aniline 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC5197-25G
PC5197-5G
PC5197-100G
APOSPC5197-25GEA
463
EUR
APOSPC5197-25G
APOSPC5197-5G
APOSPC5197-100G
2-(2,2,2-Trifluoroethoxy)aniline 97%
2-(2,2,2-Trifluoroethoxy)aniline
Formel:
C₈H₈F₃NO Flammpunkt: >110 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00077605 CAS-Nummer: 57946-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...