3-Fluor-5-methoxybenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5112-1G
PC5112-5G
APOSPC5112-1GEA
27.7
EUR
APOSPC5112-1G
APOSPC5112-5G
3-Fluor-5-methoxybenzoesäure 98%
3-Fluoro-5-methoxybenzoic acid
Formel:
C₈H₇FO₃ Schmelzpunkt: 130…133 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08235092 CAS-Nummer: 176548-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...