2-Nitro-4-(trifluormethyl)anisol 97%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-3-nitrobenzotrifluoride, 1-Methoxy-2-nitro-4-(trifluoromethyl)benzene

PC5060-100G PC5060-10G PC5060-25G PC5060-5G
APOSPC5060-100GEA 143 EUR
APOSPC5060-100G APOSPC5060-10G APOSPC5060-25G APOSPC5060-5G
2-Nitro-4-(trifluormethyl)anisol 97%
2-Nitro-4-(trifluormethyl)anisol
Formel: C₈H₆F₃NO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007099
CAS-Nummer: 394-25-2
UN-Nummer: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR