2-(Trifluoromethyl)pyrimidine-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC50209-1G PC50209-5G PC50209-25G
APOSPC50209-1GEA 50.2 EUR
APOSPC50209-1G APOSPC50209-5G APOSPC50209-25G
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
Formel: C₆H₃F₃N₂O₂
Schmelzpunkt: 170…175 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09054851
CAS-Nummer: 306960-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR