4-Fluoro-3-formylbenzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC501645-5G
PC501645-25G
APOSPC501645-5GEA
352
EUR
APOSPC501645-5G
APOSPC501645-25G
4-Fluoro-3-formylbenzoic acid 97%
4-Fluoro-3-formylbenzoic acid
Formel:
C₈H₅FO₃ Schmelzpunkt: 152…153 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06411555 CAS-Nummer: 845885-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...