Potassium (methoxymethyl)trifluoroborate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium trifluoro(methoxymethyl)borate
PC50073-1G
PC50073-5G
APOSPC50073-1GEA
26.1
EUR
APOSPC50073-1G
APOSPC50073-5G
Potassium (methoxymethyl)trifluoroborate
Potassium (methoxymethyl)trifluoroborate
Formel:
C₂H₅BF₃KO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD10566517 CAS-Nummer: 910251-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...