Kaliummethyltrifluoroborat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium trifluoro(methyl)borate, Methyltrifluoroboric acid potassium salt

PC50060-10G PC50060-5G
APOSPC50060-10GEA 104 EUR
APOSPC50060-10G APOSPC50060-5G
Kaliummethyltrifluoroborat 97%
Kaliummethyltrifluoroborat
Formel: CH₃BF₃K
Schmelzpunkt: 168…183 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08276757
CAS-Nummer: 13862-28-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR