3'-Chlor-2'-fluoracetophenon 97%

Lieferant: Apollo Scientific

Synonyme: 1-(3-Chloro-2-fluorophenyl)ethanone

PC500342-5G PC500342-1G
APOSPC500342-5GEA 79.9 EUR
APOSPC500342-5G APOSPC500342-1G
3'-Chlor-2'-fluoracetophenon 97%
3'-Chlor-2'-fluoracetophenon
Formel: C₈H₆ClFO
Molekulargewicht: 172.59 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00106165
CAS-Nummer: 161957-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR