Tetrabutylammonium bifluoride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Tetrabutylammonium Hydrogen Difluoride
PC49669-100G
PC49669-250G
PC49669-25G
APOSPC49669-100GEA
387
EUR
APOSPC49669-100G
APOSPC49669-250G
APOSPC49669-25G
Tetrabutylammonium bifluoride
Tetrabutylammonium bifluoride
Formel:
C₁₆H₃₇F₂N Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00077877 CAS-Nummer: 23868-34-0 UN-Nummer: 2924 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...