2,4-Dichlor-5-fluorbenzotrifluorid 95%
Lieferant: Apollo Scientific
PC49575-1G
PC49575-5G
PC49575-2G
APOSPC49575-1GEA
212
EUR
APOSPC49575-1G
APOSPC49575-5G
APOSPC49575-2G
2,4-Dichlor-5-fluorbenzotrifluorid 95%
2,4-Dichlor-5-fluorbenzotrifluorid
Formel:
C₇H₂Cl₂F₄ Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
112290-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...