2,3-Dichloro-5-fluoropyridine 98%

Lieferant: Apollo Scientific
Achtung

PC49213-25G PC49213-5G
APOSPC49213-25GEA 209 EUR
APOSPC49213-25G APOSPC49213-5G
2,3-Dichloro-5-fluoropyridine 98%
2,3-Dichloro-5-fluoropyridine
Formel: C₅H₂Cl₂FN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08669666
CAS-Nummer: 185985-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR