5-(Trifluormethyl)-3-pyridinboronsäurepinakolester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)pyridine
PC49119-1G
PC49119-250MG
APOSPC49119-1GEA
193
EUR
APOSPC49119-1G
APOSPC49119-250MG
5-(Trifluormethyl)-3-pyridinboronsäurepinakolester 98%
5-(Trifluormethyl)-3-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₅BF₃NO₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD12198142 CAS-Nummer: 1084953-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...