Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate

Lieferant: Apollo Scientific
Achtung

PC48337-1G PC48337-5G
APOSPC48337-1GEA 33 EUR
APOSPC48337-1G APOSPC48337-5G
Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate
Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate
Formel: C₆H₁₂BF₃KNO₂
Schmelzpunkt: 178…181 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19686142
CAS-Nummer: 1314538-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR