3'-Brom-2'-fluoracetophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Bromo-2-fluorophenyl)ethanone
PC48297-1G
PC48297-5G
PC48297-10G
APOSPC48297-1GEA
21.9
EUR
APOSPC48297-1G
APOSPC48297-5G
APOSPC48297-10G
3'-Brom-2'-fluoracetophenon 97%
3'-Brom-2'-fluoracetophenon
Formel:
C₈H₆BrFO Molekulargewicht: 217.04 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09264507 CAS-Nummer: 161957-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...