4,4,4,4',4',4'-Hexafluoro-DL-valine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4,4,4-trifluoro-3-(trifluoromethyl)butyric Acid
PC4810-1G
PC4810-250MG
APOSPC4810-1GEA
347
EUR
APOSPC4810-1G
APOSPC4810-250MG
4,4,4,4',4',4'-Hexafluoro-DL-valine 97%
4,4,4,4',4',4'-Hexafluoro-DL-valine
Formel:
C₅H₅F₆NO₂ Schmelzpunkt: 204 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014349 CAS-Nummer: 16063-80-2 EINECS: 240-207-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...