1,1,1,3,3,3-Hexafluor-2-phenyl-2-propanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,1,1,3,3,3-Hexafluoro-2-phenylpropan-2-ol, alpha,alpha-Bis(trifluoromethyl)benzyl Alcohol, 2,2,2,2',2',2'-Hexafluorocumyl Alcohol
PC4666-5G
PC4666-100G
PC4666-25G
APOSPC4666-5GEA
63.8
EUR
APOSPC4666-5G
APOSPC4666-100G
APOSPC4666-25G
1,1,1,3,3,3-Hexafluor-2-phenyl-2-propanol 97%
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol
Under Mitsunobu conditions primary and secondary alcohols can be protected as hexafluorophenylisopropyl(HIP) ethers: J. Am. Chem. Soc., 116, 8354 (1994).
Formel:
C₉H₆F₆O Siedepunkt: 159…160 °C Dichte: 1.45 g/cm³ (25 °C) Flammpunkt: 60 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040983 CAS-Nummer: 718-64-9 EINECS: 211-943-4 UN-Nummer: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...