4-(Trifluormethylthio)toluol
Lieferant: Apollo Scientific
|
Achtung
|
PC4582-1G
PC4582-250MG
PC4582-5G
APOSPC4582-1GEA
123
EUR
APOSPC4582-1G
APOSPC4582-250MG
APOSPC4582-5G
4-(Trifluormethylthio)toluol
4-(Trifluormethylthio)toluol
Formel:
C₈H₇F₃S Siedepunkt: 166…168 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00129190 CAS-Nummer: 352-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...