5-Ethoxycarbonyl-4-(trifluoromethyl)-pyrimidin-2-(1H)-one
Lieferant: Apollo Scientific
Synonyme:
Ethyl 1,2-dihydro-2-oxo-6-(trifluoromethyl)pyrimidine-5-carboxylate
PC446070-1G
PC446070-5G
APOSPC446070-1GEA
29.3
EUR
APOSPC446070-1G
APOSPC446070-5G
5-Ethoxycarbonyl-4-(trifluoromethyl)-pyrimidin-2-(1H)-one
5-Ethoxycarbonyl-4-(trifluoromethyl)-pyrimidin-2-(1H)-one
Formel:
C₈H₇F₃N₂O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00052077 CAS-Nummer: 154934-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...