2-Amino-3-nitro-5-fluorpyridin 98%

Lieferant: Apollo Scientific

PC445028-1G PC445028-5G
APOSPC445028-1GEA 23 EUR
APOSPC445028-1G APOSPC445028-5G
2-Amino-3-nitro-5-fluorpyridin 98%
2-Amino-3-nitro-5-fluorpyridin
Formel: C₅H₄FN₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04972413
CAS-Nummer: 212268-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR