5-Fluoro-2-iodopyridine 97%
Lieferant: Apollo Scientific
Synonyme:
2-Iodo-5-fluoropyridine
PC445023-250MG
PC445023-1G
APOSPC445023-250MGEA
87.3
EUR
APOSPC445023-250MG
APOSPC445023-1G
5-Fluoro-2-iodopyridine 97%
5-Fluoro-2-iodopyridine
Formel:
C5H3FIN Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07787574 CAS-Nummer: 159870-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...