(Perfluoroheptanoyl)acetone 97%

Lieferant: Apollo Scientific

PC4436-5G PC4436-1G PC4436-25G
APOSPC4436-5GEA 137 EUR
APOSPC4436-5G APOSPC4436-1G APOSPC4436-25G
(Perfluoroheptanoyl)acetone 97%
(Perfluoroheptanoyl)acetone
Formel: C₁₀H₅F₁₃O₂
Molekulargewicht: 404.13 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04038336
CAS-Nummer: 82822-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR