4-Fluor-2-(trifluormethyl)benzylamin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(4-Fluoro-2-(trifluoromethyl)phenyl)methanamine
PC4374Y-25G
PC4374Y-5G
PC4374Y-100G
APOSPC4374Y-25GEA
137
EUR
APOSPC4374Y-25G
APOSPC4374Y-5G
APOSPC4374Y-100G
4-Fluor-2-(trifluormethyl)benzylamin 97%
4-Fluor-2-(trifluormethyl)benzylamin
Formel:
C₈H₇F₄N Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00061164 CAS-Nummer: 202522-22-3 UN-Nummer: 2735 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...