α,α,α,2-Tetrafluor-p-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-(trifluoromethyl)benzoic acid
PC4373-100G
PC4373-25G
PC4373-5G
APOSPC4373-100GEA
306
EUR
APOSPC4373-100G
APOSPC4373-25G
APOSPC4373-5G
α,α,α,2-Tetrafluor-p-toluylsäure 98%
α,α,α,2-Tetrafluor-p-toluylsäure
Formel:
C₈H₄F₄O₂ Schmelzpunkt: 166…170 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010322 CAS-Nummer: 115029-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...