α,α,α,2-Tetrafluor-m-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4372T-100G
PC4372T-25G
PC4372T-5G
APOSPC4372T-100GEA
179
EUR
APOSPC4372T-100G
APOSPC4372T-25G
APOSPC4372T-5G
α,α,α,2-Tetrafluor-m-toluylsäure 98%
α,α,α,2-Tetrafluor-m-toluylsäure
Formel:
C₈H₄F₄O₂ Schmelzpunkt: 126…128 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00040980 CAS-Nummer: 115029-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...