4-Fluor-2-(trifluormethyl)benzamid 97%
Lieferant: Apollo Scientific
PC4371W-25G
PC4371W-5G
PC4371W-1G
APOSPC4371W-25GEA
200
EUR
APOSPC4371W-25G
APOSPC4371W-5G
APOSPC4371W-1G
4-Fluor-2-(trifluormethyl)benzamid 97%
4-Fluor-2-(trifluormethyl)benzamid
Formel:
C₈H₅F₄NO Schmelzpunkt: 134…137 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00061150 CAS-Nummer: 207919-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...