Kalium(1-methyl-4-trifluoroboratomethyl)piperazin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium trifluoro((4-methylpiperazin-1-yl)methyl)borate

PC430511-1G PC430511-25G PC430511-5G
APOSPC430511-1GEA 37.7 EUR
APOSPC430511-1G APOSPC430511-25G APOSPC430511-5G
Kalium(1-methyl-4-trifluoroboratomethyl)piperazin 95%
Kalium(1-methyl-4-trifluoroboratomethyl)piperazin
Formel: C₆H₁₃BF₃KN₂
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD10700160
CAS-Nummer: 1015484-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR