Kalium(morpholin-4-ylmethyl)trifluoroborat
Lieferant: Apollo Scientific
Synonyme:
Potassium trifluoro(morpholinomethyl)borate
PC430508-1G
PC430508-5G
APOSPC430508-1GEA
111
EUR
APOSPC430508-1G
APOSPC430508-5G
Kalium(morpholin-4-ylmethyl)trifluoroborat
Kalium(morpholin-4-ylmethyl)trifluoroborat
Formel:
C₅H₁₀BF₃KNO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD10566512 CAS-Nummer: 936329-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...