3-Chlor-3'-fluorbenzhydrol

Lieferant: Apollo Scientific

PC4220-1G PC4220-250MG
APOSPC4220-1GEA 691 EUR
APOSPC4220-1G APOSPC4220-250MG
3-Chlor-3'-fluorbenzhydrol
3-Chlor-3'-fluorbenzhydrol
Formel: C₁₃H₁₀ClFO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06201311
CAS-Nummer: 170019-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR