3-Nitro-4-(trifluoromethoxy)benzoic acid 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC408277-25G
PC408277-5G
PC408277-100G
APOSPC408277-25GEA
43.5
EUR
APOSPC408277-25G
APOSPC408277-5G
APOSPC408277-100G
3-Nitro-4-(trifluoromethoxy)benzoic acid 99%
3-Nitro-4-(trifluoromethoxy)benzoic acid
Formel:
C₈H₄F₃NO₅ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13185757 CAS-Nummer: 784-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...