3-Fluor-L-phenylalanin

Lieferant: Apollo Scientific
Achtung

PC4025-5G PC4025-500MG
APOSPC4025-5GEA 143 EUR
APOSPC4025-5G APOSPC4025-500MG
3-Fluor-L-phenylalanin
3-Fluor-L-phenylalanin
Formel: C₉H₁₀FNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00066450
CAS-Nummer: 19883-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR