Fmoc-4-(trifluoromethyl)-D-phenylalanine

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-Phe(4-CF3)-OH, Fmoc-D-Phe(4-CF₃)-OH

PC4013-1G PC4013-5G
APOSPC4013-1GEA 121 EUR
APOSPC4013-1G APOSPC4013-5G
Fmoc-4-(trifluoromethyl)-D-phenylalanine
Fmoc-4-(trifluoromethyl)-D-phenylalanine
Formel: C₂₅H₂₀F₃NO₄
Molekulargewicht: 455.44 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00797582
CAS-Nummer: 238742-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR