5-Brom-2-hydroxy-4-(trifluormethyl)pyridin

Lieferant: Apollo Scientific
Achtung

PC401004-1G PC401004-5G
APOSPC401004-1GEA 69.6 EUR
APOSPC401004-1G APOSPC401004-5G
5-Brom-2-hydroxy-4-(trifluormethyl)pyridin
5-Brom-2-hydroxy-4-(trifluormethyl)pyridin
Formel: C₆H₃BrF₃NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11227189
CAS-Nummer: 109919-32-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR