3-Brom-5-(trifluormethoxy)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC3956-10G PC3956-5G
APOSPC3956-10GEA 328 EUR
APOSPC3956-10G APOSPC3956-5G
3-Brom-5-(trifluormethoxy)benzylalkohol 98%
3-Brom-5-(trifluormethoxy)benzylalkohol
Formel: C₈H₆BrF₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09999512
CAS-Nummer: 1026201-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR