2-Amino-4-t-butyl-6-(trifluoromethyl)pyrimidine 97%
Lieferant: Apollo Scientific
PC3881-1G
PC3881-250MG
APOSPC3881-1GEA
130
EUR
APOSPC3881-1G
APOSPC3881-250MG
2-Amino-4-t-butyl-6-(trifluoromethyl)pyrimidine 97%
2-Amino-4-t-butyl-6-(trifluoromethyl)pyrimidine
Formel:
C₉H₁₂F₃N₃ Schmelzpunkt: 48…50 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00458223 CAS-Nummer: 238742-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...