Poly(chlortrifluorethylen) 800
Lieferant: Apollo Scientific
PC3814D-1LB
PC3814D-5LB
APOSPC3814D-1LBEA
1020
EUR
APOSPC3814D-1LB
APOSPC3814D-5LB
Poly(chlortrifluorethylen) 800
Poly(chlortrifluorethylen) 800
Formel:
(C₂ClF₃)n Dichte: 1,9 g/cm³ (25 °C) Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
9002-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...