4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-2-(methylsulphanyl)-6-(trifluoromethyl)pyrimidine
PC3648-1G
PC3648-5G
APOSPC3648-1GEA
24.8
EUR
APOSPC3648-1G
APOSPC3648-5G
4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine
4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine
Formel:
C₆H₄ClF₃N₂S Molekulargewicht: 228.617 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00269080 CAS-Nummer: 16097-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...