5-Carbamoyl-2-fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3573-1G
PC3573-5G
APOSPC3573-1GEA
178
EUR
APOSPC3573-1G
APOSPC3573-5G
5-Carbamoyl-2-fluorphenylboronsäure 97%
5-Carbamoyl-2-fluorphenylboronsäure
Formel:
C₇H₇BFNO₃ Schmelzpunkt: 192…194 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08436055 CAS-Nummer: 874289-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...