DL-2-Fluoro-3-alanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-2-fluoropropanoic acid
PC3387E-250MG
PC3387E-100MG
APOSPC3387E-250MGEA
337
EUR
APOSPC3387E-250MG
APOSPC3387E-100MG
DL-2-Fluoro-3-alanine
DL-2-Fluoro-3-alanine
Formel:
C₃H₆FNO₂ Schmelzpunkt: 222…223 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00041324 CAS-Nummer: 3821-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...